Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRANDplates immunoGrade™ 96-Well Microplates

Catalog No. 13882218 Shop All BrandTech Scientific Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Clear
White
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round
Round, C Bottom
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Well Shape
13-882-218 Clear Round
13-882-219 Clear Conical Bottom (V-well)
13-882-220 Clear Flat Bottom (F-well)
13-882-222 White Round
13-882-226 Black, Clear Bottom Flat Bottom (F-well)
13-882-223 White Round, C Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 13-882-218 Supplier BRANDTECH™ Supplier No. 781720
Add to Cart
Add to Cart

An excellent choice for the majority of standard ELISAs as well as solid phase immunoassays

  • Optimized for the immobilization of IgG
  • High binding capacity for molecules with mixed hydrophobic and hydrophilic regions
  • Designed to minimize cross-contamination
  • Blue alphanumeric embossing for easy well identification on standard plates
  • Non-sterile

Order Info

Packaging is 20 stacks of 5 plates each (100 plates). Each stack comes with one lid.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type High Binding
Color Clear
Product Type 96-well Microplate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment immunoGrade
Total Volume (Metric) Well 330 μL
Well Shape Round
For Use With (Application) For the majority of standard ELISAs as well as solid phase immunoassays
Quantity 100/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.