Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ BRANDplates™ immunoGrade™ 384-Well Microplates

Catalog No. 14386032 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Color:
Black
Clear
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-386-032 Black
14-386-030 Clear
14-386-031 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-386-032 Supplier BRANDTECH™ BRAND™ Supplier No. 781742
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Immunoassay plate optimized for immobilization of IgG

  • High binding for molecules with mixed hydrophilic and hydrophobic regions
  • Surface choice for majority of standard ELISAs
  • For solid phase immunoassays
  • PS, non-sterile
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase, non-cytotoxic
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Lid With Lid
Array 16 x 24
Color Black
Material Polystyrene
Volume (Metric) Well 100 μL
No. of Wells 384
Sterility Non-sterile
Surface Treatment immunoGrade
Well Shape Flat Bottom (F-well)
For Use With (Application) For immobilization of IgG, solid phase immunoassays
Quantity 50/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.