Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ 1536-Well Microplates

Catalog No. 14386005 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Color:
Black
Clear
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-386-005 Black
14-386-003 Clear
14-386-004 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-386-005 Supplier BRANDTECH™ BRAND™ Supplier No. 781642
Add to Cart
Add to Cart

Non-treated microplates for diverse applications

  • Nontreated PS surface
  • Medium binding, nonsterile
  • Standard plate for most applications
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic

Homogenous assays, screening, and storage.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Medium Binding
Certifications/Compliance ISO 11137, AAMI
Format Standard
Lid Without Lid
Array 32 x 48
Autoclavable Not Autoclavable
Binding Type Untreated
Color Black
Disposable Single-use
Product Type Detection Plate
Skirt Style Full
Material Polystyrene
Volume (Metric) Well 10 μL
No. of Wells 1536
Volume (Metric) Working 10 μL
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 10.536 mL
Well Shape Flat Bottom (F-well)
For Use With (Application) Sterile applications, bacteriological assays
Quantity 50/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.