Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ BRANDplates™ pureGrade™ S 96-well Microplates

Catalog No. p-4830454 Shop All BrandTech Scientific Products
Change view
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
White
White, Clear Bottom
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round Bottom (U-well)
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Well Shape
13-882-210 Clear Round Bottom (U-well)
13-882-211 Clear Conical Bottom (V-well)
13-882-212 Clear Flat Bottom (F-well)
13-882-214 White Flat Bottom (F-well)
13-882-215 Black Flat Bottom (F-well)
13-882-216 White, Clear Bottom Flat Bottom (F-well)
13-882-217 Black, Clear Bottom Flat Bottom (F-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 13-882-210 Supplier BRANDTECH™ BRAND™ Supplier No. 781660
Only null left
Add to Cart
Edge
Add to Cart

Nontreated sterile microplates for diverse applications

  • PS surface
  • Medium binding
  • Sterilized via beta-radiation
  • Standard plates available in U-, V-, F-, C-shapes
  • Transparent bottom available in F- shape
  • With lid
  • Available in clear, white or black
  • Gray alphanumeric embossing on white and black solid plates for easy well identification
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic

Bacteriological assays, screenings, storage.

TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Certifications/Compliance ISO 11137, AAMI, DIN EN ISO 10993, ANSI, SLAS
Format Standard
Lid With Lid
Array 8 x 12
Binding Type Medium Binding
Color Clear
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Sterile
Surface Treatment Non-treated
Well Shape Round Bottom (U-well)
For Use With (Application) Bacteriological assays, screening and for storage
Quantity 50/Cs.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.