Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Shelf/Rack mount for HandyStep™ S

Catalog No. 13689213 Shop All BrandTech Scientific Products
Change view
Click to view available options
Positions:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
13-689-213 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-689-213 Supplier BRANDTECH™ BRAND™ Supplier No. 705130
Add to Cart
Add to Cart

Pipetter Shelf/Rack Mount for HandyStep™ S Electronic Repetitive Pipet

  • For use with HandyStep™ S Repetitive Pipet
TRUSTED_SUSTAINABILITY

Specifications

Description Shelf/rack mount
Autoclavable Not Autoclavable
Color Gray
Holds Manual Pipettes
Material Plastic
Positions 1
Shape Circular
Stackable No
Sterility Non-sterile
For Use With (Equipment) HandyStep S Electronic
Product Type Pipette Holder
Shelves None
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.