Learn More
BRANDTECH™ BRAND™ inertGrade™ 96-Well Non-Binding Microplates, PS
Catalog No. 14386220 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Color:
Transparent
Catalog No. Color
14-386-220 Transparent
1 options
This item is not returnable. View return policy
Catalog No. 14-386-220 Supplier BRANDTECH™ BRAND™ Supplier No. 781900
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sterile inertGrade™ treated microplates for the cultivation of non-adherent cell lines and suspension cultures.
  • Designed for cultivating suspension cell lines where adhesion is undesirable
  • Optimized surface reduces cell adhesion and protein adsorption, minimizing enzyme and cellular activation
  • Inhibits early differentiation of stem cells
  • Orange color code with embossed alphanumeric coding for easy identification
  • Made from pure, newly synthesized polystyrene (PS), sterile, and includes lids
  • BIO-CERT™ CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, pyrogens, and non-cytotoxic
  • Manufactured in an ISO Class 7 cleanroom for optimal cleanliness
  • Raw materials meet USP and ISO 10993 requirements
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Array 8 x 12
Autoclavable Not Autoclavable
Packaging Individually wrapped
Color Transparent
Product Type 96-well microplate
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Non-binding
Well Shape Round Bottom (U-well)
No. per Case 40/Cs.
No. per Pack 40
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov