Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ PLT™ Pipette Leak Testing Unit

Catalog No. 03837733 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
03-837-733 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-837-733 Supplier BRANDTECH™ BRAND™ Supplier No. 703970
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Improves process reliability by detecting the smallest leaks in air displacement pipettes.

The BRAND PLT™ Pipet Leak Tester quickly and easily detects even the smallest leaks.

  • Identifies leaks invisible to the naked eye within seconds.
  • Pre-programmed for all single- and multichannel pipettes from 1 μL to 10 mL.
  • Test with or without tips.
  • Choose between dynamic or static testing.
TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips All tips
Autoclavable Not Autoclavable
Color Gray
Disposable Reusable
For Use With Pipettes
Material Plastic
Number of Channels Variable
Programmability No
Sterility Non-sterile
Type Leak Tester
For Use With (Equipment) Air displacement pipettes from all manufacturers
Includes Adapter, 2 plugs, 3 replacement polyethylene filters, universal AC adapter, quality certificate, operating manual
Electrical Requirements 100/200V, 50/60Hz
Voltage 100/200 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.