Learn More
BRANDTECH™ VACUUBRAND™ Quick Coupling for BVC Fluid Aspiration Systems
Catalog No. p-4879303 Shop All BrandTech Scientific Products
Change view
Click to view available options
For Use With (Application):
Collection Vessel/Pump Coupling
VHC/Collection Vessel Coupling
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
13-688-021 VHC/Collection Vessel Coupling
13-688-022 Collection Vessel/Pump Coupling
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-688-021 Supplier BRANDTECH™ VACUUBRAND™ Supplier No. 20635807
Only null left
Add to Cart
Add to Cart
Replacement parts and accessories for VACUUBRAND BVC fluid aspiration systems
The BVC family of fluid aspiration systems deliver compact and quiet solutions for cell culture work. They provide the flexibility to accommodate decontamination protocol whether autoclaving or disinfecting in the bottle.
635807
  • Quick coupling from VHC to collection vessel
635808
  • Quick coupling from collection vessel to pump
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) VHC/Collection Vessel Coupling
For Use With BVC fluid aspiration system
Type Quick Coupling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov