Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VITLAB™ Reagent Bottles with Stoppers

Catalog No. 14379106 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
5000 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14-379-106 1000 mL
14-379-103 100 mL
14-379-104 250 mL
14-379-105 500 mL
14-379-108 5000 mL
14-379-107 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-379-106 Supplier BRANDTECH™ VITLAB™ Supplier No. 100694
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VITLAB polypropylene reagent bottles with polypropylene stoppers have good chemical resistance. Autoclavable at 121 C for 20 minutes.

  • NS stoppers and good chemical resistance for long-term storage of liquids.
  • Autoclavable at 121 C, according to DIN EN 285
TRUSTED_SUSTAINABILITY

Specifications

Capacity 33.8 oz., 1000mL
Capacity (Metric) 1000 mL
Color Transparent
Diameter (English) 4.25 in.
Material Polypropylene
Quantity 10/Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 213 mm
Height (English) 8.38 in.
Closure Material Polypropylene
Closure Size 29/32 mm
Diameter (Metric) 108 mm
Autoclavable Autoclavable
Capacity (English) 33.8 oz.
Product Type Reagent Bottle
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.