Learn More
BRANDTECH™ BRAND™ Battery Pack for HandyStep™ touch Repeating Pipette
GSA_VA
Catalog No. 13688403 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Battery Type:
NiMH
Catalog No. Battery Type
13-688-403 NiMH
1 options
Catalog No. 13-688-403 Supplier BRANDTECH™ BRAND™ Supplier No. 705025
Add to Cart
Edge
Add to Cart
Replacement battery pack for use with HandyStep™ touch and HandyStep™ touch S Repeating Pipette.
  • Replaceable nickel-metal-hydride battery pack charges in 2.5 hours (3.5 hours inside instrument)
  • Battery pack can be easily interchanged for continuous pipetting
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Battery Type NiMH
For Use With BRAND HandyStep Touch/Touch S
Material Plastic
Type Battery Pack
For Use With (Equipment) HandyStep electronic repeating pipette battery
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov