Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Centrifuge Tube Mini-Cooler

Catalog No. 14380802 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Color:
Red
White
Yellow
Holds Temp., Hours:
-20°C for 60 min.
-70°C for 45 min.
0°C for 60 min.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Holds Temp., Hours
14-380-802 White -70°C for 45 min.
14-380-800 Red 0°C for 60 min.
14-380-801 Yellow -20°C for 60 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-380-802 Supplier BRANDTECH™ BRAND™ Supplier No. 114940
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench.

  • Manufactured from durable polycarbonate
  • Filled with non-toxic gel
  • Hold twelve 0.5 to 2.0 mL tubes
  • Provided individually
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Centrifuge tube
Holds Temp., Hours -70°C for 45 min.
Material Polycarbonate
Product Type Mini Cooler
Color White
Capacity (Metric) 0.5 to 2 mL
Holds 0.5 to 2 mL Tubes
Autoclavable Not Autoclavable
Temperature Control Passive
Temperature Range -70°C
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.