Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Transferpette™ Electronic Single Channel Pipette AC Adapter

Catalog No. 13689215 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Plug Type:
US
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type
13-689-215 US
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-689-215 Supplier BRANDTECH™ BRAND™ Supplier No. 705352
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Power supply for Transferpette™ electronic pipettes, US plug

  • For single and multichannel models
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Black
Disposable Reusable
For Use With Transferpette Electronic Single Channel Pipette
Material HDPE
Number of Channels Variable
Sterility Non-sterile
Type Power Supply
For Use With (Equipment) Transferpette electronic pipette
Plug Type US
Voltage 110 V
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.