Learn More
BRANDTECH™ VACUUBRAND™ Oil-Free Chemistry Design Diaphragm Pump Key
Catalog No. 13688978 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Overall Width (English):
2.59 in.
Catalog No. Overall Width (English)
13-688-978 2.59 in.
1 options
Catalog No. 13-688-978 Supplier BRANDTECH™ VACUUBRAND™ Supplier No. 20636554
Add to Cart
Edge
Add to Cart
For diaphragm pumps NT
  • Diaphragm key (wrench) to replace diaphragms on VACUUBRAND 'NT' model vacuum pumps
TRUSTED_SUSTAINABILITY
Specifications
For Use With Oil free diaphragm vacuum pump
Overall Width (English) 2.59 in.
Type Diaphragm Key
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov