Learn More
BRANDTECH™ VACUUBRAND™ Rotary Vacuum Pumps
Catalog No. 1387860 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
VACUUBRAND™ Rotary Vacuum Pumps
Catalog No.
13-878-60 VACUUBRAND™ Rotary Vacuum Pumps
1 options
Catalog No. 13-878-60 Supplier BRANDTECH™ Supplier No. 697051
Includes
All models are supplied oil filled with ON/OFF control switch, cable with plug, small flange centring and clamping rings for inlet and outlet, and instructions.
Compact, oil-sealed, rotary vane vacuum pumps. Choose between one- and two-stage pumps according to ultimate vacuum required. An outlet-based anti-suckback valve keeps the pump chamber under vacuum. This reduces condensation of corrosive vapors inside the pump if the pump is switched off. Integrated gas ballast for use with vapors that can condense inside the pump. Large oil volume for longer intervals between oil changes. High splash protection and overload circuit breaker for safety. Start-up power control for easy start-up at low ambient temperature and less energy consumption (not available in RE2). Vanes and seal materials are asbestos free.
Order Info
For 120V 60Hz. For other voltages, contact your Customer Service Representative.