Learn More
BRANDTECH™ VACUUBRAND™ Service Kits for MZ 2C Vacuum Pumps
Catalog No. 13878118 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Type:
Service Kit
Catalog No. Type
13-878-118 Service Kit
1 options
Catalog No. 13-878-118 Supplier BRANDTECH™ VACUUBRAND™ Supplier No. 20696814
Add to Cart
Add to Cart
Service kits for VACUUBRAND™ Chemistry Diaphragm Pumps.
  • Sets include appropriate diaphragms, valves and sealing rings (if applicable)
  • Wrench not included.
TRUSTED_SUSTAINABILITY
Specifications
For Use With Model ME4C and MZ2C vacuum pumps
Includes 2 Diaphragms and 4 valves
Type Service Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov