Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VACUUBRAND™ VACUUBUS™ Vacuum Controls and Gauges Extension Cable

Catalog No. 13688963 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Overall Length (Metric):
200 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Overall Length (Metric)
13-688-963 200 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-688-963 Supplier BRANDTECH™ VACUUBRAND™ Supplier No. 612552

missing translation for 'validMainframeMsg'

For use with DCP3000, CVC3000 and other VACUU·BUS compliant products

  • Length: 2m
TRUSTED_SUSTAINABILITY

Specifications

For Use With CVC3000 and DCP3000 + VSK3000 vacuum controls and gauges
Overall Length (English) 78.74 in.
Overall Length (Metric) 200 cm
Type Extension Cable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.