Learn More
BRANDTECH™ VACUUBRAND™ XS Series Rotary Vane Vacuum Pumps
Catalog No. 13889135 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Plug Type:
US
Catalog No. Plug Type
13-889-135 US
1 options
Catalog No. 13-889-135 Supplier BRANDTECH™ Supplier No. 20698126
Designed for very low noise and vibration.
  • Small and energy efficient: Substantially smaller and lighter than pumps of comparable capacity, saving you bench or floor space. Motors were selected for their energy efficiency.
  • Rugged and low maintenance: Features many nitrogen-plasma treated parts in flowpath to increase corrosion resistance. Integrated gas ballast helps ensure exceptional water vapor tolerance with minimal impact on ultimate vacuum. Wearing forces have also been reduced for longer service interval
  • For applications where diaphragm pumps cannot provide sufficient vacuum, small freeze dryers, small centrifugal concentrators or other low-temperature evaporations of high boiling point solvents, molecular distillations, and other applications where diaphragm pumps cannot provide sufficient vacuum
TRUSTED_SUSTAINABILITY
Specifications
Plug Type US
Pump Type Rotary Vane Vacuum Pump
Applications Applications where diaphragm pumps cannot provide sufficient vacuum
Voltage 120 V