Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VITLAB™ Polypropylene Screw Caps

Catalog No. 14379307 Shop All BrandTech Scientific Products
Change view
Click to view available options
Thread Size (Metric):
GL 18
GL 32
GL 45
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
14-379-307 GL 18
14-379-309 GL 32
14-379-311 GL 45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-379-307 Supplier BRANDTECH™ VITLAB™ Supplier No. 83310
Add to Cart
Add to Cart

Screw caps for VITLAB™ and other GL threaded bottles

  • Polypropylene construction
  • Transparent
  • Autoclavable at 121°C
TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Material Polypropylene
Sterility Non-sterile
Thread Size (Metric) GL 18
Disposable Limited-use
Product Type Screw Cap
For Use With (Equipment) Vitlab and other GL threaded bottle
Autoclavable Autoclavable
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.