Learn More
BRANDTECH™ VITLAB™ SAN Tall Form Graduated Cylinders, Class B
Shop All BRANDTECH Scientific ProductsDescription
- Made of crystal-clear Styrene Acrylonitrile (SAN).
- Hexagonal base with bottom studs provides high stability and resists rolling.
- Thermal stress up to 60°C does not cause tolerance limits to be permanently exceeded.
Specifications
Specifications
| Class | Class B |
| Quantity | 6 Pk. |
| Sterility | Non-sterile |
| Base Type | Hexagonal |
| Product Type | Graduated Cylinder |