Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ VITLAB™ Wide-Mouth Reagent Bottles with Screw Caps

Catalog No. 14379109 Shop All BrandTech Scientific Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Size:
GL 32
GL 45
GL 63
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size
14-379-109 100 mL GL 32
14-379-353 250 mL GL 45
14-379-354 500 mL GL 45
14-379-112 1000 mL GL 63
14-379-113 2000 mL GL 63
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-379-109 Supplier BRANDTECH™ VITLAB™ Supplier No. 101589
Add to Cart
Add to Cart

VITLAB wide mouth polypropylene reagent bottles with polypropylene screw caps have good chemical resistance. Autoclavable at 121 C for 20 minutes.

  • Wide mouth for easier access to contents
  • Screw caps help protect agaist outside contamination and leakage
TRUSTED_SUSTAINABILITY

Specifications

Capacity 3.38 oz., 100 mL
Capacity (Metric) 100 mL
Capacity (English) 3.38 oz.
Color Transparent
Diameter (English) 2.16 in.
Material Polypropylene
Quantity 20/Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide
Height (Metric) 96 mm
Product Type Reagent Bottle
Height (English) 3.77 in.
Closure Material Polypropylene
Closure Size GL 32
Diameter (Metric) 55 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.