Learn More
BRANDTECH™ VITLAB™ Wide-Mouth Reagent Bottles with Screw Caps
Shop All BRANDTECH Scientific ProductsDescription
- Wide mouth for easier access to contents
- Screw caps help protect agaist outside contamination and leakage
Specifications
Specifications
| Capacity | 33.8 oz., 1000 mL |
| Capacity (Metric) | 1000 mL |
| Color | Transparent |
| Diameter (English) | 4.25 in. |
| Material | Polypropylene |
| Quantity | 10/Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.