Learn More
Medchemexpress LLC HY-112161 5mg , Branebrutinib CAS:1912445-55-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1121615MG
Encompass_Preferred
HY-112161 5mg Branebrutinib CAS: 1912445-55-6 Branebrutinib (BMS-986195) is a highly potent, selective covalent, irreversible inhibitor of Bruton’s tyrosine kinase (BTK), with an IC50 of 0.1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0546