Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Branson Ultrasonics™ Bransonic™ Metal Cleaners

Catalog No. 1533726 Shop All Branson Ultrasonics Corporation Products
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
Product Type:
MC3 Metal Cleaner
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Product Type
15-337-26 5 gal. MC3 Metal Cleaner
15-335-52 1 gal. MC3 Metal Cleaner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-337-26 Supplier Branson Ultrasonics™ Supplier No. 100955846

missing translation for 'validMainframeMsg'

For ultrasonic cleaners

Specifications

Product Type MC3 Metal Cleaner
Form Liquid
For Use With (Application) Used to remove Oils, Greases and soils from Aluminum and Aluminum alloys, Copper, Brass and Steel substrates
Includes MC-3 Metal Cleaner
Volume (English) 5 gal.
Volume (Metric) 19 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.