Learn More
Sigma Aldrich Fine Chemicals Biosciences Branson CPX series ultraso1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7695091EA
Encompass
With the Bransonic(R) CPX weve gone digital and defined a new category in ultrasonic baths. Weve reimagined our iconic line of ultrasonic baths for total control with a fully digital Tower Control Panel. The CPX also features our new self-adaptive technology that actively tracks and adapts to changes within the bath.All Bransonic(R) baths can also be used for cell separation sample preparation and degassing of liquids.FeaturesDegassing - can be set up to 99 minutesTimer - can be set to 99 minutes or run continuouslySelf-Adaptive technologyDigital tower control panelSleep modeAll 220-230 V units meet CE standardsNote Parts should not be allowed to rest on tank bottom see accessories for options.
Catalog No. 50-290-8649