Learn More
Sigma Aldrich Fine Chemicals Biosciences Branson CPXH series ultras1EA
Catalog No. 502908992
Encompass
The Bransonic(R) CPXH digital bath is our most advanced ultrasonic bath yet. It features the benefits of heated cleaning with the ability to set temperatures from 20C/68F - 69C/156.2F. Plus you get complete digital control with programmable capabilities and self-adaptive technology for consistent worry-free operation - every time.All Bransonic(R) baths can also be used for cell separation sample preparation and degassing of liquids.FeaturesDegassing - can be set up to 99 minutesTimer - can be set to 99 minutes or run continuouslyHeated Cleaning - up to 69C/156.2FSelf-Adaptive TechnologyFully ProgrammableHigh/low Power ControlSleep ModeCSA compliantCE compliantNote Parts should not be allowed to rest on tank bottom see accessories for options.
Catalog No. 50-290-8992 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7692071EA
The Bransonic(R) CPXH digital bath is our most advanced ultrasonic bath yet. It features the benefits of heated cleaning with the ability to set temperatures from 20C/68F - 69C/156.2F. Plus you get complete digital control with programmable capabilities and self-adaptive technology for consistent worry-free operation - every time.All Bransonic(R) baths can also be used for cell separation sample preparation and degassing of liquids.FeaturesDegassing - can be set up to 99 minutesTimer - can be set to 99 minutes or run continuouslyHeated Cleaning - up to 69C/156.2FSelf-Adaptive TechnologyFully ProgrammableHigh/low Power ControlSleep ModeCSA compliantCE compliantNote Parts should not be allowed to rest on tank bottom see accessories for options.