Learn More
Sigma Aldrich Fine Chemicals Biosciences Branson CPXH series ultras1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7693471EA
Encompass
The Bransonic(R) CPXH digital bath is our most advanced ultrasonic bath yet. It features the benefits of heated cleaning with the ability to set temperatures from 20C/68F - 69C/156.2F. Plus you get complete digital control with programmable capabilities and self-adaptive technology for consistent worry-free operation - every time.All Bransonic(R) baths can also be used for cell separation sample preparation and degassing of liquids.Features.nbspDegassing - can be set up to 99 minutes.nbspTimer - can be set to 99 minutes or run continuously.nbspHeated Cleaning - up to 69C/156.2F.nbspSelf-Adaptive Technology.nbspFully Programmable.nbspHigh/low Power Control.nbspSleep Mode.nbspCSA compliant.nbspCE compliantNote Parts should not be allowed to rest on tank bottom see accessories for options.
Catalog No. 50-290-8555