Learn More
Sigma Aldrich Fine Chemicals Biosciences Branson M series ultrasoni1EA
Catalog No. 502908554
Encompass
The Bransonic(R) M Ultrasonic Cleaning Bath is the leading mechanical ultrasonic bath. Its designed for performance control durability and reliability for a variety of applications.All Bransonic(R) baths can also be used for cell separation sample preparation and degassing of liquids.Features40kHz Industrial Transducers60-minute Mechanical Timer (can be set to run continuously)Tower Control PanelBuilt-in Pour Drain (A½ A¾ gallon) or Side Mounted DrainNote Parts should not be allowed to rest on tank bottom see accessories for options.
Catalog No. 50-290-8554 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7695761EA
The Bransonic(R) M Ultrasonic Cleaning Bath is the leading mechanical ultrasonic bath. Its designed for performance control durability and reliability for a variety of applications.All Bransonic(R) baths can also be used for cell separation sample preparation and degassing of liquids.Features40kHz Industrial Transducers60-minute Mechanical Timer (can be set to run continuously)Tower Control PanelBuilt-in Pour Drain (A½ A¾ gallon) or Side Mounted DrainNote Parts should not be allowed to rest on tank bottom see accessories for options.