Learn More
Sigma Aldrich Fine Chemicals Biosciences Branson MH series ultrason1EA
Catalog No. 502908609
Encompass
With the Bransonic(R) MH Ultrasonic bath you get the added benefit of heat coupled with the durability and performance of our flagship ultrasonic bath. It also features a temperature sensor and auto shut-off when the unit reaches 69C/156.2F.All Bransonic(R) baths can also be used for cell separation sample preparation and degassing of liquids.FeaturesHeated Cleaning - up to 69C/156.2F60-minute Mechanical Timer (can be set to run continuously)40kHz Industrial TransducersTower Control PanelBuilt-in Pour Drain (A½ A¾ gallon) or Side Mounted DrainAll ultrasonic baths meet FCC CSA CE UL rules and regulationsNote Parts should not be allowed to rest on tank bottom see accessories for options.
Catalog No. 50-290-8609 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7697461EA
With the Bransonic(R) MH Ultrasonic bath you get the added benefit of heat coupled with the durability and performance of our flagship ultrasonic bath. It also features a temperature sensor and auto shut-off when the unit reaches 69C/156.2F.All Bransonic(R) baths can also be used for cell separation sample preparation and degassing of liquids.FeaturesHeated Cleaning - up to 69C/156.2F60-minute Mechanical Timer (can be set to run continuously)40kHz Industrial TransducersTower Control PanelBuilt-in Pour Drain (A½ A¾ gallon) or Side Mounted DrainAll ultrasonic baths meet FCC CSA CE UL rules and regulationsNote Parts should not be allowed to rest on tank bottom see accessories for options.