Learn More
Electron Microscopy Sciences Branson Ultra Sonic Bath Model 8800-MT, 115 V
SDP
Catalog No. NC1546318
Encompass_Preferred
With a unique sweep frequency technology to eliminate standing waves, & a pioneering 40 kHz ind. transducers, Bransonic ultrasonic baths have been the industry std for quality, reliability, & precision cleaning.
Constant activity/power tracking automatically maintains the ultrasonic power by adjusting for changes in liquid level & temp caused by a light or heavy load. This helps ensure more uniform cleaning & consistent performance.
High/low power control adjusts the acoustic energy in the bath & can be set at 100% power for normal to heavier loads or at 70% power for lighter, This helps to protect components from potential damage.
temp settings up to 69°C/156.2°F can be set in C or F & programmed through the front panel for easy access & monitoring. High-temp visual alarm/auto shut-off provides safety. Ultrasonic Cleaner only w/ mechanical timer, tank size: 19.5"x11"x6", tank capacity: 5.5 gal, drain: yes, overall size: 24"x18"x14.5".
Catalog No. NC1546318 Supplier Electron Microscopy Sciences Supplier No. 71040MT
With a unique sweep frequency technology to eliminate standing waves, & a pioneering 40 kHz ind. transducers, Bransonic ultrasonic baths have been the industry std for quality, reliability, & precision cleaning.
Constant activity/power tracking automatically maintains the ultrasonic power by adjusting for changes in liquid level & temp caused by a light or heavy load. This helps ensure more uniform cleaning & consistent performance.
High/low power control adjusts the acoustic energy in the bath & can be set at 100% power for normal to heavier loads or at 70% power for lighter, This helps to protect components from potential damage.
temp settings up to 69°C/156.2°F can be set in C or F & programmed through the front panel for easy access & monitoring. High-temp visual alarm/auto shut-off provides safety. Ultrasonic Cleaner only w/ mechanical timer, tank size: 19.5"x11"x6", tank capacity: 5.5 gal, drain: yes, overall size: 24"x18"x14.5".