Learn More
Branson Ultrasonics™ Sonifier Spanner Wrench
Catalog No. 22309794 Shop All Branson Ultrasonics Corporation Products
Change view
Click to view available options
For Use With (Equipment):
250 and 450 Sonifier Cell Disruptor
Catalog No. For Use With (Equipment)
22-309794 250 and 450 Sonifier Cell Disruptor
1 options
Catalog No. 22-309794 Supplier Branson Ultrasonics™ Supplier No. 101118039
Add to Cart
Add to Cart
Requires
2 wrenches needed; one for converter and one for probe
Removes disruptor from 250 and 450 model Sonifiers™
Removes disruptor from 250 and 450 model Sonifiers™
Made from titanium
2 required; One for converter and one for probe
Specifications
Material Titanium
For Use With (Equipment) 250 and 450 Sonifier Cell Disruptor
Product Type Wrench Spanner