Learn More
Electron Microscopy Sciences Brasil Fixative 10 ML
SDP
Catalog No. 501901164
Encompass_Preferred
Ethanol, Picric Acid, Formalin, Acetic Acid, Glacial. This fixative is most popular when glycogen is the target. The solution is similar to Bouins Solution (alcoholic) offering quick hardening and good staining with acid dyes. In 20 ml glass vials.
Catalog No. 50-190-1164 Supplier Electron Microscopy Sciences Supplier No. 1599510
Ethanol, Picric Acid, Formalin, Acetic Acid, Glacial. This fixative is most popular when glycogen is the target. The solution is similar to Bouins Solution (alcoholic) offering quick hardening and good staining with acid dyes. In 20 ml glass vials.