Learn More
Cayman Chemical BrasInazole 10mg
SDP
Catalog No. 502980285
Encompass_Preferred
An inhibitor of brassinosteroid biosynthesis that at 1 µM can induce morphological changes, including dwarfism and altered leaf color and curling, in dark-grown Arabidopsis and light-grown cress
Catalog No. 50-298-0285 Supplier Cayman Chemical Supplier No. 1680410 MG
An inhibitor of brassinosteroid biosynthesis that at 1 µM can induce morphological changes, including dwarfism and altered leaf color and curling, in dark-grown Arabidopsis and light-grown cress