Learn More
Cayman Chemical Brassinazole, Purity: ≥98%, CAS: 224047-41-0, 50 mg
SDP
Catalog No. 502979851
Encompass_Preferred
An inhibitor of brassinosteroid biosynthesis that at 1 µM can induce morphological changes, including dwarfism and altered leaf color and curling, in dark-grown Arabidopsis and light-grown cress
Catalog No. 50-297-9851 Supplier Cayman Chemical Supplier No. 1680450 MG
An inhibitor of brassinosteroid biosynthesis that at 1 µM can induce morphological changes, including dwarfism and altered leaf color and curling, in dark-grown Arabidopsis and light-grown cress