Learn More
Cayman Chemical BrasInolIde 10mg
SDP
Supplier:  Cayman Chemical 2159410 MG
Encompass_Preferred
A biologically active brassinosteroid that has important roles in regulating plant growth and development
Catalog No. 50-298-5916