Learn More
Electron Microscopy Sciences Wood Handle Brass Bristle Brush
SDP
Catalog No. 5031578
Encompass_Preferred
A four-row brush made in Germany from 0.10 mm crimped steel or brass wire set in a wooden handle. Excellent for use as a cleaning brush or for surface texturing on soft metals. The overall length is 8 5/8" .
Catalog No. 50-315-78 Supplier Electron Microscopy Sciences Supplier No. 65585B
A four-row brush made in Germany from 0.10 mm crimped steel or brass wire set in a wooden handle. Excellent for use as a cleaning brush or for surface texturing on soft metals. The overall length is 8 5/8" .