Learn More
Icl Cal Lab Inc A70 Brass Cage Sampler, 32oz
SDP
Catalog No. NC1375455
Encompass
For sampling from tank cars, tank trucks, shore tanks, ship tanks and barge tanks. Takes all level samples, running samples, and top, upper, middle, lower and outlet samples.
Catalog No. NC1375455 Supplier Icl Cal Lab Inc Supplier No. A70QB
For sampling from tank cars, tank trucks, shore tanks, ship tanks and barge tanks. Takes all level samples, running samples, and top, upper, middle, lower and outlet samples.