Learn More
Perkin Elmer US LLC Brass Hex Nipple, 0.25 in NPT
Catalog No. 502112692
Encompass_Preferred
This brass male connector is included in Installation Kit - Clarus (N6500123). Style: Hex Nipple Connection 1: 1/4 inch Male NPT Connection 2: 1/4 inch Male NPT Hex: 9/16 inch Pipe Thread: 1/4 inch Flow Diameter: 0.314 inch
Catalog No. 50-211-2692 Supplier Perkin Elmer US LLC Supplier No. 09200213
This brass male connector is included in Installation Kit - Clarus (N6500123). Style: Hex Nipple Connection 1: 1/4 inch Male NPT Connection 2: 1/4 inch Male NPT Hex: 9/16 inch Pipe Thread: 1/4 inch Flow Diameter: 0.314 inch