Learn More
Grainger ASC Engineered Solutions Inline Check Valve, Lead Free Brass, 3/4 in. NPT, 200 psi, 1 ea.
Catalog No. 502785616
Encompass_Preferred
This in-line check valve is designed for reliable flow control in various piping applications. It features a lead-free brass construction, ensuring compliance with drinking water regulations, and is equipped with a spring-loaded mechanism to prevent backflow efficiently.
  • Lead-free brass body construction
  • Features an EPDM seat and seal
  • Includes a stainless steel spring
  • Operates at a pressure rating of 200 psi
  • Opens at a cracking pressure of 2 psi
  • Designed with threaded NPT end connections
  • Certified to NSF/ANSI 372 standards
  • Approved for California AB1953 lead-free requirements
Catalog No. 50-278-5616 Supplier Grainger Supplier No. 195L51
This in-line check valve is designed for reliable flow control in various piping applications. It features a lead-free brass construction, ensuring compliance with drinking water regulations, and is equipped with a spring-loaded mechanism to prevent backflow efficiently.
  • Lead-free brass body construction
  • Features an EPDM seat and seal
  • Includes a stainless steel spring
  • Operates at a pressure rating of 200 psi
  • Opens at a cracking pressure of 2 psi
  • Designed with threaded NPT end connections
  • Certified to NSF/ANSI 372 standards
  • Approved for California AB1953 lead-free requirements