Learn More
Waters Corp Waters Corporation Seat, Brass, 515 Pump, 100 μL Configuration
Catalog No. 50793510 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Corporation brass seat is a critical component for 515 Pumps configured for 100 μL operation. It functions as a plunger seat, ensuring proper sealing and performance within the pump system. This component is essential for maintaining the specified performance and can be re-used when converting the pump back to its original 100 μL configuration after installing an Extended Flow Range Kit.
  • Constructed from brass.
  • Designed as a plunger seat.
  • Essential for 100 μL pump configurations.
  • Used in the 515 Pump.
  • Component for converting pump back to 100 μL setup.
Catalog No. 50-793-510 Supplier Waters Corp Supplier No. WAT023060
The Waters Corporation brass seat is a critical component for 515 Pumps configured for 100 μL operation. It functions as a plunger seat, ensuring proper sealing and performance within the pump system. This component is essential for maintaining the specified performance and can be re-used when converting the pump back to its original 100 μL configuration after installing an Extended Flow Range Kit.
  • Constructed from brass.
  • Designed as a plunger seat.
  • Essential for 100 μL pump configurations.
  • Used in the 515 Pump.
  • Component for converting pump back to 100 μL setup.