Learn More
Glass Expansion Brass Skimmer Adaptor Base for Agilent 7900 X Lens (G8400-60625)
SDP
Catalog No. 502703000
Encompass_Preferred
Brass Skimmer Adaptor Base for Agilent 7900 X Lens (G8400-60625). For use with platinum skimmer cone on Agilent 7900 ICP-MS with x-lens.Replacement Captive Screws M4 for Brass Skimmer Adaptor Base for Agilent 7900 X Lens are 70-803-2061.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3000 Supplier Glass Expansion Supplier No. AT7905XBR
Brass Skimmer Adaptor Base for Agilent 7900 X Lens (G8400-60625). For use with platinum skimmer cone on Agilent 7900 ICP-MS with x-lens.Replacement Captive Screws M4 for Brass Skimmer Adaptor Base for Agilent 7900 X Lens are 70-803-2061.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001