Learn More
Grainger Central Tools Slide Rule Caliper, Brass, 0-100 mm, 1 ea.
Catalog No. 502788172
Encompass_Preferred
This slide rule caliper is constructed from heavy gauge solid brass and finished in satin chrome for durability. It features a fine line indicator hand and is designed for versatile measurement tasks in the shop or laboratory.
  • Measures inside, outside, depth, and step dimensions
  • Range of 0 to 4 in. (0 to 100 mm)
  • Graduations of 1/32 in. and 1 mm
Catalog No. 50-278-8172 Supplier Grainger Supplier No. 168Z58
This slide rule caliper is constructed from heavy gauge solid brass and finished in satin chrome for durability. It features a fine line indicator hand and is designed for versatile measurement tasks in the shop or laboratory.
  • Measures inside, outside, depth, and step dimensions
  • Range of 0 to 4 in. (0 to 100 mm)
  • Graduations of 1/32 in. and 1 mm