Learn More
MSC Precision Brand Feeler Gage, 0.009 in. Thick, CDA 260 Alloy Brass, 10/Pk.
Catalog No. 5000514259
Encompass
These brass thickness gages are precision-engineered for use in manufacturing and maintenance operations that require checking extremely tight tolerances or close spacing. Constructed from 260 alloy brass, these nonmagnetic and noncorrosive blades are designed to be spark-resistant, making them ideal for specialized applications in the computer, mining, and electronics industries.
  • Made from 1/2 in. wide 260 brass alloy
  • Measures 5 in. long with a 0.009 in. thickness
  • Nonmagnetic and noncorrosive properties for safe use in sensitive environments
  • Spark-resistant material suitable for hazardous area applications
  • Thickness clearly etched every 6 in. in metric and decimal
  • Includes 10 blades per package
Catalog No. 50-005-14259 Supplier MSC Supplier No. 79010963
These brass thickness gages are precision-engineered for use in manufacturing and maintenance operations that require checking extremely tight tolerances or close spacing. Constructed from 260 alloy brass, these nonmagnetic and noncorrosive blades are designed to be spark-resistant, making them ideal for specialized applications in the computer, mining, and electronics industries.
  • Made from 1/2 in. wide 260 brass alloy
  • Measures 5 in. long with a 0.009 in. thickness
  • Nonmagnetic and noncorrosive properties for safe use in sensitive environments
  • Spark-resistant material suitable for hazardous area applications
  • Thickness clearly etched every 6 in. in metric and decimal
  • Includes 10 blades per package