Learn More
Apexbio Technology LLC Brassinolide 72962-43-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064379
Encompass_Preferred
Brassinolide (CAS 72962-43-7) is a naturally occurring steroidal plant growth regulator It influences various physiological processes in plants including stem elongation leaf differentiation flower formation and fruit development In vitro studies indicated that brassinolide induces apoptosis in human prostate cancer PC-3 cells characterized by increased caspase-3 activity and decreased Bcl-2 protein expression In animal models brassinolide reduced blood glucose levels in diabetic rats demonstrating non-dose-dependent hypoglycemic effects without observed toxicity Brassinolide is valuable in research related to cancer cell apoptosis and glucose metabolism regulation
Catalog No. 50-000-64379 Supplier Apexbio Technology LLC Supplier No. A32655.1
Brassinolide (CAS 72962-43-7) is a naturally occurring steroidal plant growth regulator It influences various physiological processes in plants including stem elongation leaf differentiation flower formation and fruit development In vitro studies indicated that brassinolide induces apoptosis in human prostate cancer PC-3 cells characterized by increased caspase-3 activity and decreased Bcl-2 protein expression In animal models brassinolide reduced blood glucose levels in diabetic rats demonstrating non-dose-dependent hypoglycemic effects without observed toxicity Brassinolide is valuable in research related to cancer cell apoptosis and glucose metabolism regulation