Learn More
Apexbio Technology LLC Brassinolide, 72962-43-7, MFCD00792745, 5 mg
SDP
Supplier:  Apexbio Technology LLC A32655
Encompass_Preferred
MF: C28H48O6, MW: 480.68, Solubility: 48.1 mg/mL in DMSO with gentle warming, 52.3 mg/mL in EtOH with ultrasonic and warming, insoluble in H2O. Brassinolide is a plant growth regulator. Plant growth regulators are naturally produced by plants and are critical for regulating their own growth. Plant growth regulators act via modifying or controlling plant growth processes, such as leave and flower formation, stem elongation, as well as fruit development and ripening.
Catalog No. 50-115-0534