Learn More
Aviva Systems Biology Corp BRAT1 Antibody - C-terminal region (ARP62762_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62762_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BRAT1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human BRAT1. Alternate names: MGC22916, BAAT1, C7orf27ApplicationWB. Species reactivity: Human
Catalog No. 50-287-764