Learn More
McKesson General Medical BRAUN 414995 CASE/50
Catalog No. NC1498147
Encompass_Preferred
Low drug binding membrane, proximal and distal Luer lock connections designed for fungal and particulate filtration of medication. Fluid retention: 0.3 mL, Color: Clear
This item is not returnable. View return policy
Catalog No. NC1498147 Supplier McKesson General Medical Supplier No. 239659
Low drug binding membrane, proximal and distal Luer lock connections designed for fungal and particulate filtration of medication. Fluid retention: 0.3 mL, Color: Clear