Learn More
Med Vet International Braun Cassette #2/0 USP 100m (328')L. Medium, J0091020, Each
Catalog No. 5000582427
Encompass_Preferred
Braunamid. Suture consists of a large number of fine twisted strands covered by a smooth, seamless, homogeneous polyamide cover. Because of its polyfil construction, Braunamid thread is considerably more flexible than the monofil sutures. Knot-pull tensile strength substantially exceeds USP requirements. Knot-holding ability is excellent due to its inner polyfilament structure. Non-capillary. Sterilized. Braunamid meets or exceeds all legal requirements set forth by the U.S. government.
Catalog No. 50-005-82427 Supplier Med Vet International Supplier No. JOR91020
Braunamid. Suture consists of a large number of fine twisted strands covered by a smooth, seamless, homogeneous polyamide cover. Because of its polyfil construction, Braunamid thread is considerably more flexible than the monofil sutures. Knot-pull tensile strength substantially exceeds USP requirements. Knot-holding ability is excellent due to its inner polyfilament structure. Non-capillary. Sterilized. Braunamid meets or exceeds all legal requirements set forth by the U.S. government.