Learn More
Med Vet International Braun Cassette #4/0 4/0 USP 100m(328')L.Medium Fine, J0091040, Each
Catalog No. 5000581162
Encompass_Preferred
Braunamid. Suture consists of a large number of fine twisted strands covered by a smooth, seamless, homogeneous polyamide cover. Because of its polyfil construction, Braunamid thread is considerably more flexible than the monofil sutures. Knot-pull tensil
Catalog No. 50-005-81162 Supplier Med Vet International Supplier No. JOR91040
Braunamid. Suture consists of a large number of fine twisted strands covered by a smooth, seamless, homogeneous polyamide cover. Because of its polyfil construction, Braunamid thread is considerably more flexible than the monofil sutures. Knot-pull tensil