Learn More
Med Vet International Braun Cassette #8 8 USP 25m (82')L. Special, J009108, Each
Catalog No. 5000582527
Encompass_Preferred
Braunamid. Suture consists of a large number of fine twisted strands covered by a smooth, seamless, homogeneous polyamide cover. Because of its polyfil construction, Braunamid thread is considerably more flexible than the monofil sutures. Knot-pull tensil
Catalog No. 50-005-82527 Supplier Med Vet International Supplier No. JOR9108
Braunamid. Suture consists of a large number of fine twisted strands covered by a smooth, seamless, homogeneous polyamide cover. Because of its polyfil construction, Braunamid thread is considerably more flexible than the monofil sutures. Knot-pull tensil