Learn More
Med Vet International Braun Cassette Suture, #6 6 USP 25m (82')L., J009106, Each
Catalog No. 5000582472
Encompass_Preferred
Braunamid. Suture consists of a large number of fine twisted strands covered by a smooth, seamless, homogeneous polyamide cover. Because of its polyfil construction, Braunamid thread is considerably more flexible than the monofil sutures. Knot-pull tensil
Catalog No. 50-005-82472 Supplier Med Vet International Supplier No. JOR9106
Braunamid. Suture consists of a large number of fine twisted strands covered by a smooth, seamless, homogeneous polyamide cover. Because of its polyfil construction, Braunamid thread is considerably more flexible than the monofil sutures. Knot-pull tensil